Iron(II) chloride tetrahydrate, EMSURE® for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

M-Clarity™ quality level = MQ300

1.03861.0250 1.03861.1000 1.03861.9050
1.03861.0250EA 82.7 EUR
1.03861.0250 1.03861.1000 1.03861.9050
Iron(II) chloride tetrahydrate, EMSURE® for analysis, Supelco®
Iron(II) chloride tetrahydrate
Formule: FeCl₂·4H₂O
MW: 198.81 g/mol
Kookpunt: 1,023 °C
Smeltpunt: 105…110 °C (–H₂O van kristall.)
Dichtheid: 1.936 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149709
CAS nummer: 13478-10-9
EINECS: 231-843-4
UN: 3260

Order Now

Specification Test Results

Assay (manganometric) ≥ 98.0 %
Sulfate (SO₄) ≤ 0.01 %
Total nitrogen (N) ≤ 0.001 %
As (Arsenic) ≤ 0.0005 %
Cu (Copper) ≤ 0.002 %
Mn (Manganese) ≤ 0.005 %
Pb (Lead) ≤ 0.001 %
Zn (Zinc) ≤ 0.003 %
Substances not precipitated by ammonia (as sulfates) ≤ 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR