Formamide, EMPLURA®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Carbamaldehyde

M-Clarity™ quality level = MQ200

1.04008.9025
1.04008.9025EA 0 EUR
1.04008.9025
Formamide, EMPLURA®, Supelco®
Formamide
Formule: HCONH₂
Kookpunt: 210 °C (1013 hPa)
Smeltpunt: 2 °C
Dichtheid: 1,1334 g/cm³ (20 °C)
Vlampunt: 79 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007941
CAS nummer: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR