n-Heptaan, EMPLURA®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

1.04365.1000 1.04365.2500 1.04365.6025 1.04365.6190 1.04365.9950 1.04365.2511 1.04365.9011
1.04365.1000EA 62.9 EUR
1.04365.1000 1.04365.2500 1.04365.6025 1.04365.6190 1.04365.9950 1.04365.2511 1.04365.9011
n-Heptaan, EMPLURA®, Supelco®
n-Heptaan

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formule: H₃C(CH₂)₅CH₃
Kookpunt: 98,4 °C (1013 hPa)
Dichtheid: 0,6838 g/cm³ (20 °C)
Vlampunt: –4 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009544
CAS nummer: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR