2,2,4-Trimethylpentane, Uvasol® Spectroscopy, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

MQ100

1.04718.0500 1.04718.2500
1.04718.0500EA 122 EUR
1.04718.0500 1.04718.2500
2,2,4-Trimethylpentane, Uvasol® Spectroscopy, Supelco®
2,2,4-Trimethylpentane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formule: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Kookpunt: 99.2
Smeltpunt: –107 °C
Dichtheid: 0.692 g/cm³ (20 °C)
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥99.8%
Evaporation Residue ≤0.0002%
Water ≤0.005%
Color ≤10 Hazen
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Fluorescence (as quinine at 254 nm) ≤1.0 ppb
Fluorescence (as quinine at 365 nm) ≤1.0 ppb
Transmission (at 205 nm) ≥30%
Transmission (at 215 nm) ≥65%
Transmission (at 220 nm) ≥80%
Transmission (at 225 nm) ≥85%
Transmission (at 235 nm) ≥90%
Transmission (at 245 nm) ≥98%
Transmission (from 255 nm) ≥99%
Absorbance (at 205 nm) ≤0.52
Absorbance (at 215 nm) ≤0.19
Absorbance (at 220 nm) ≤0.10
Absorbance (at 225 nm) ≤0.07
Absorbance (at 235 nm) ≤0.05
Absorbance (at 245 nm) ≤0.01
Absorbance (from 255 nm) ≤0.004
Meet the requirements of 'Specific use spectroscopy' according to ACS and Reag. Ph. Eur.

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR