2,2,4-Trimethylpentane, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

MQ300

1.04727.1000 1.04727.2500 1.04727.6010 1.04727.6025 1.04727.6190 1.04727.4000
1.04727.1000EA 49.05 EUR
1.04727.1000 1.04727.2500 1.04727.6010 1.04727.6025 1.04727.6190 1.04727.4000
2,2,4-Trimethylpentane, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
2,2,4-Trimethylpentane

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formule: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Kookpunt: 99.2
Smeltpunt: –107 °C
Dichtheid: 0.692 g/cm³ (20 °C)
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.56 mg/l (25 °C)
Melting Point -107 °C
Molar Mass 114.23 g/mol
Boiling Point 99 °C (1013 hPa)
Vapor Pressure 51 hPa (20 °C)
Flash Point -12 °C
Refractive Index 2.3
Explosion Limit 1 %(V)
Density 0.69 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 410 °C
Saturation concentration 239 g/m3 (20 °C)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR