Jood Reag. Ph. Eur., ACS, ISO zA, sublimed, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


MQ100

1.04761.0100 1.04761.0500
1.04761.0100EA 93.6 EUR
1.04761.0100 1.04761.0500
Jood Reag. Ph. Eur., ACS, ISO zA, sublimed, Supelco®
Jood

Certistain dyes for microscopy are tested for their chemical composition and in particular for their functional use in accordance with strict specifications.


  • The dyes ensure consistent staining results.

Formule: I₂
MW: 253.81 g/mol
Kookpunt: 184.3
Smeltpunt: 114
Dichtheid: 4.9 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011355
CAS nummer: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 (6.1) III
Merck Index: 13,05036

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 114 °C
Molar Mass 253.8 g/mol
Bulk Density 2100 kg/m3
Boiling Point 185 °C (1013 hPa)
Vapor Pressure 0.41 hPa (25 °C)
Density 4.93 g/cm3 (20 °C)
pH 5.4 (H2O) (saturated solution)
Viscosity Kinematic 0.57 mm2/s (116 °C) liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR