Potassium acetate BP, Ph. Eur., E261, extra_pure, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid potassium salt

1.04820.1000 1.04820.5000 1.04820.9025 1.04820.9050
1.04820.1000EA 161 EUR
1.04820.1000 1.04820.5000 1.04820.9025 1.04820.9050
Potassium acetate BP, Ph. Eur., E261, extra_pure, SAFC®
Potassium acetate
Formule: CH₃COOK
MW: 98.14 g/mol
Smeltpunt: 304
Dichtheid: 1.57 g/cm³ (25 °C)
Vlampunt: >250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012458
CAS nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR