Potassium chloride, EMSURE® for analysis, Supelco®

Leverancier: Merck
Ratings:
Avg. Ratings: 4.0 (1 Review)

Total Ratings: 1
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

M-Clarity™ quality level = MQ300

1.04936.0500 1.04936.1000 1.04936.5000 1.04936.9050 1.04936.9010 1.04936.0250
1.04936.0500EA 30.29 EUR
1.04936.0500 1.04936.1000 1.04936.5000 1.04936.9050 1.04936.9010 1.04936.0250
Potassium chloride, EMSURE® for analysis, Supelco®
Potassium chloride
Formule: KCl
Kookpunt: 1420 °C (1013 hPa)
Smeltpunt: 773 °C
Dichtheid: 1,98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Solubility 347 g/l (20 °C)
Melting Point 773 °C
Molar Mass 74.55 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1413 °C (1013 hPa)
Density 1.98 g/cm3 (20 °C)
pH 5.5 - 8.0 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR