Kaliumhydroxide, pellets, pure, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potash caustic, Caustic potash

M-Clarity™ quality level = MQ200

1.05012.1000 1.05012.5000 1.05012.9050
1.05012.1000EA 56.9 EUR
1.05012.1000 1.05012.5000 1.05012.9050
Kaliumhydroxide, pellets, pure, Supelco®
Kaliumhydroxide
Formule: KOH
Kookpunt: 1320 °C (1013 hPa)
Smeltpunt: 360 °C
Dichtheid: 2,04 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
Bereik: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Solubility 1130 g/l (20 °C)
Melting Point 360 °C
Molar Mass 56.11 g/mol
Boiling Point 1320 °C
Vapor Pressure (20 °C)
Density 2.04 g/cm3 (20 °C)
pH 14 (56 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR