Potassium iodide ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ300

1.05043.0250 1.05043.0500 1.05043.1000 1.05043.2500 1.05043.9050
1.05043.0250EA 214 EUR
1.05043.0250 1.05043.0500 1.05043.1000 1.05043.2500 1.05043.9050
Potassium iodide ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Potassium iodide
Formule: Kl
Kookpunt: 1330 °C (1013 hPa)
Smeltpunt: 686 °C
Dichtheid: 3,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011405
CAS nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Solubility 1430 g/l (20 °C)
Melting Point 723 °C
Molar Mass 166.00 g/mol
Bulk Density 1500 kg/m3
Boiling Point 1325 °C (1013 hPa)
Vapor Pressure 1.3 hPa (745 °C)
Density 3.13 g/cm3 (20 °C)
pH 6.9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR