Potassium thiocyanate, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potassium rhodanide

M-Clarity™ quality level = MQ200

1.05124.1000 1.05124.9050
1.05124.1000EA 83.5 EUR
1.05124.1000 1.05124.9050
Potassium thiocyanate, Supelco®
Potassium thiocyanate
Formule: KSCN
Kookpunt: 500 °C (1013 hPa)
Smeltpunt: 175 °C
Dichtheid: 1,61 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011413
CAS nummer: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR