Eusolex® 2292

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.05382.0101 1.05382.1001 1.05382.9026 1.05382.0100
1.05382.0101EA 0 EUR
1.05382.0101 1.05382.1001 1.05382.9026 1.05382.0100
Eusolex® 2292
Eusolex®

Organic UV filter


Eusolex® 2292 is a clear, liquid, oil soluble and effective UV-B filter. It's easy to process in an oil phase and shows good solubilizing properties for solid crystalline UV filters. Eusolex® 2292 is worldwide approved.


INCI name: Ethylhexyl Methoxycinnamate, BHT

Formule: C₁₈H₂₆O₃
Kookpunt: 160 °C (1 hPa)
Dichtheid: 1.005…1.013 g/cm3 (25 °C)
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 5466-77-3

Order Now

Specification Test Results

Color colourless to light yellow (< 100 Hazen)
Appearance clear liquid
Odour practical odourless
Assay Ethylhexyl methoxycinnamate (GC) 95.0 - 105.0 %
Chromatography purity conforms
Identity (retention time) conforms
Identit (IR-spectrum) conforms
Refractive index (n 20/D) 1.542 - 1.548
Density (d 20/20) 1.010 - 1.016
Density (d 25/25) 1.005 - 1.013
Spee. absorbance(at 308 nm; 1 %; 1 cm; methanol) ≥ 830
Heavy metals (as Pb) ≤ 0.002%
As (Arsenic) ≤ 0.0002%
Residual Solvents ICHQ3C excluded by manufacturing process
Acidity conforms
Sulfated ash ≤ 0.1 %
Water ≤ 0.5 %
Microbial Purity (Total Viable Aerobic Count) ≤ 100 CFU/g
E.coli absent in 1 g
Pseudomonas aeruginosa absent in 1 g
Staphylococcus aureus absent in 1 g
Candida albicans absent in 1 g
Salmonella species absent in 10 g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR