Ammoniak-oplossing 25%, Suprapur® for trace analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ammoniumhydroxide, Waterig-ammoniakale oplossing

M-Clarity™ quality level = MQ100

1.05428.0250 1.05428.1000 1.05428.0500 1.05428.2500
1.05428.0250EA 126 EUR
1.05428.0250 1.05428.1000 1.05428.0500 1.05428.2500
Ammoniak-oplossing 25%, Suprapur® for trace analysis, Supelco®
Ammoniak-oplossing
Formule: NH₃
MW: 35.05 g/mol
Kookpunt: 32…37.7 °C (1013 hPa)
Smeltpunt: –60…–57.5 °C
Dichtheid: 0.88…0.91 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066650
CAS nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -57.5 °C
Boiling Point 37.7 °C
Vapor Pressure 483 hPa (20 °C)
Explosion Limit 15.4 - 33.6 %(V)
Density 0.903 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly alkaline
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR