Zinc iodide and starch solution analytical reagent, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

1.05445.0500
1.05445.0500EA 54.1 EUR
1.05445.0500
Zinc iodide and starch solution analytical reagent, Supelco®
Zinc iodide starch solution

Order Now

Specification Test Results

Solubility Soluble at 20 °C
Density 1.02 g/cm3 (20 °C)
pH 6.2 (H2O, 20 °C)
Storage temperature +15...+25 °C
Suitability as indicator conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR