Lithium nitrate, Suprapur® for trace analysis, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
M-Clarity™ quality level = MQ100
1.05653.0100 1.05653.0500
1.05653.0100EA 502 EUR
1.05653.0100 1.05653.0500
Lithium nitrate, Suprapur® for trace analysis, Supelco®
Lithium nitrate
Formule: LiNO₃
Smeltpunt: 255 °C
Dichtheid: 2,38 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011094
CAS nummer: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 13,05558
Order Now
Specification Test Results
Solubility 522 g/l (20 °C)
Melting Point 255 °C
Molar Mass 68.94 g/mol
Bulk Density 910 kg/m3
Density 2.36 g/cm3 (20 °C)
pH 7 - 9 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer