Methanol, SupraSolv® GC, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

MQ100

1.06011.1000 1.06011.2500 1.06011.4000
1.06011.1000EA 63.3 EUR
1.06011.1000 1.06011.2500 1.06011.4000
Methanol, SupraSolv® GC, Supelco®
Methanol
Formule: CH₃OH
MW: 32.04 g/mol
Kookpunt: 64.6
Smeltpunt: –98 °C
Dichtheid: 0.7918 g/cm³ (20 °C)
Vlampunt: 11 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
Bereik: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -98 °C
Molar Mass 32.04 g/mol
Boiling Point 64.5 °C (1013 hPa)
Vapor Pressure 128 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.33
Explosion Limit 5.5 - 44 %(V)
Density 0.792 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 455 °C DIN 51794
Evaporation number 6.3
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR