Methyleenchloride stabilised, LiChrosolv® for HPLC, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylene chloride

Stabilisatie: Stabilised with 2-methyl-2-butene (iso-amylene) 50 ppm
M-Clarity™ quality level = MQ100

1.06044.1000 1.06044.2500 1.06044.4000 1.06044.9010 1.06044.9030
1.06044.1000EA 95.1 EUR
1.06044.1000 1.06044.2500 1.06044.4000 1.06044.9010 1.06044.9030
Methyleenchloride stabilised, LiChrosolv® for HPLC, Supelco®
Methyleenchloride

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.

Formule: CH₂Cl₂
Kookpunt: 39,8 °C (1013 hPa)
Smeltpunt: –95 °C
Dichtheid: 1,322 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point -95 °C
Molar Mass 84.93 g/mol
Boiling Point 40 °C (1013 hPa)
Vapor Pressure 475 hPa (20 °C)
Refractive Index 1.42
Explosion Limit 13 - 22 %(V)
Density 1.33 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 605 °C DIN 51794
Synonyms DCM, Methylene chloride, Methylene dichloride
Hill Formula CH2Cl2
Chemical Formula CH2Cl2
LD50 oral LD50 Rat 1600 mg/kg
LD50 dermal LD50 Rat greater than 2000 mg/kg
Storage Store at +15°C to +25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR