N-(1-Naphthyl)ethylenediamine dihydrochloride ACS analytical reagent, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

M-Clarity™ quality level = MQ100

1.06237.0005 1.06237.0025
1.06237.0005EA 89 EUR
1.06237.0005 1.06237.0025
N-(1-Naphthyl)ethylenediamine dihydrochloride ACS analytical reagent, Supelco®
N-(1-Naphthyl)ethylenediamine dihydrochloride
Formule: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Smeltpunt: 188…190 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012556
CAS nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 30 g/l (20 °C)
Molar Mass 259.18 g/mol
Bulk Density 380 kg/m3
pH 1.0 (25 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR