Natriumacetaat, anhydrous, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid sodium salt

M-Clarity™ quality level = MQ300

1.06268.0250 1.06268.1000 1.06268.2500 1.06268.9025 1.06268.9012
1.06268.0250EA 46.6 EUR
1.06268.0250 1.06268.1000 1.06268.2500 1.06268.9025 1.06268.9012
Natriumacetaat, anhydrous, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Natriumacetaat
Formule: H₃CCOONa
MW: 82.03 g/mol
Kookpunt: 881.4
Smeltpunt: 324
Dichtheid: 1.528 g/cm³ (20 °C)
Vlampunt: >250 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012459
CAS nummer: 127-09-3
EINECS: 204-823-8
Bereik: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Solubility 365 g/l (20 °C)
Melting Point 324 °C (decomposition)
Molar Mass 82.04 g/mol
Boiling Point 400 °C (decomposition)
Vapor Pressure (20 °C)
Flash Point 250 °C
Density 1.52 g/cm3 (20 °C)
pH 7.5 - 9.0 (50 g/l, H2O, 20 °C)
Ignition Point 607 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR