Natrium-diwaterstoffosfaat-monohydraat, EMPROVE® EXPERT USP, BP, extra pure suitable for use as excipient, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

1.06349.1000 1.06349.9025 1.06349.9014
1.06349.1000EA 149 EUR
1.06349.1000 1.06349.9025 1.06349.9014
Natrium-diwaterstoffosfaat-monohydraat, EMPROVE® EXPERT USP, BP, extra pure suitable for use as excipient, SAFC®
Natrium-diwaterstoffosfaat-monohydraat
Formule: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Smeltpunt: 100
Dichtheid: 2.04 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149208
CAS nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR