Sodium carbonate decahydrate, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Soda

M-Clarity™ quality level = MQ300

1.06391.1000 1.06391.5000 1.06391.9025
1.06391.1000EA 50.68 EUR
1.06391.1000 1.06391.5000 1.06391.9025
Sodium carbonate decahydrate, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®
Sodium carbonate decahydrate
Formule: Na₂CO₃·10H₂O
MW: 286.14 g/mol
Smeltpunt: 33
Dichtheid: 1.46 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149178
CAS nummer: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Solubility 210 g/l (20 °C)
Melting Point 33 °C Elimination of water of crystallisation
Molar Mass 286.14 g/mol
Bulk Density 700 - 900 kg/m3
Density 1.44 g/cm3 (20 °C)
pH 11 - 12 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR