tri-Sodium citrate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.06447.5000 1.06447.9029
1.06447.5000EA 588 EUR
1.06447.5000 1.06447.9029
tri-Sodium citrate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®
tri-Sodium citrate dihydrate
Formule: Na₃C₆H₅O₇·2H₂O
Smeltpunt: 150 °C
Dichtheid: 1,76 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150031
CAS nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR