Natriumthiosufaat, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

1.06512.2500 1.06512.9050 1.06512.0250 1.06512.9025
1.06512.2500EA 82.3 EUR
1.06512.2500 1.06512.9050 1.06512.0250 1.06512.9025
Natriumthiosufaat, Supelco®
Natriumthiosufaat
Formule: Na₂S₂O₃
Kookpunt: 100 °C (1013 hPa)
Smeltpunt: 48 °C
Dichtheid: 1,66 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR