Sodium nitroprusside dihydrate ACS, Reag. Ph. Eur. analytical reagent, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

M-Clarity™ quality level = MQ100

1.06541.0025 1.06541.0100
1.06541.0025EA 91.6 EUR
1.06541.0025 1.06541.0100
Sodium nitroprusside dihydrate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Sodium nitroprusside dihydrate
Formule: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Dichtheid: 1.72 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149192
CAS nummer: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1 II
Merck Index: 13,08722

Order Now

Specification Test Results

Solubility 400 g/l (20 °C) (slow decomposition)
Molar Mass 297.95 g/mol
Bulk Density 1000 kg/m3
Density 1.71 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR