Sodium perchlorate monohydrate, EMSURE® for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Perchloric acid sodium salt

M-Clarity™ quality level = MQ300

1.06564.0100 1.06564.0500 1.06564.9025
1.06564.0100EA 81.1 EUR
1.06564.0100 1.06564.0500 1.06564.9025
Sodium perchlorate monohydrate, EMSURE® for analysis, Supelco®
Sodium perchlorate monohydrate
Formule: NaClO₄·H₂O
Smeltpunt: 130 °C
Dichtheid: 2,02 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149164
CAS nummer: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798

Order Now

Specification Test Results

Assay (alkalimetric) 99.0 - 102.0 %
pH-value (5 %; water) 4.5 - 7.0
Chloride (Cl) ≤ 0.002 %
Chloride, Chlorate (as Cl) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.002 %
Total nitrogen (N) ≤ 0.0005 %
Ca (Calcium) ≤ 0.002 %
Fe (Iron) ≤ 0.0003 %
K (Potassium) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR