Disodium hydrogen phosphate dodecahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
1.06573.5000EA 372 EUR
1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
Disodium hydrogen phosphate dodecahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Disodium hydrogen phosphate dodecahydrate
Formule: Na₂HPO₄·12H₂O
Smeltpunt: 35 °C
Dichtheid: 1,52 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00149181
CAS nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR