Disodium hydrogen phosphate, watervrij ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

1.06586.0500 1.06586.2500 1.06586.9050 1.06586.1000 1.06586.9012
1.06586.0500EA 46.17 EUR
1.06586.0500 1.06586.2500 1.06586.9050 1.06586.1000 1.06586.9012
Disodium hydrogen phosphate, watervrij ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Disodium hydrogen phosphate

Certificaten: Corresponds to ACS, Reag. Ph Eur

Formule: Na₂HPO₄
Smeltpunt: 250 °C
Dichtheid: 1,53 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003496
CAS nummer: 7558-79-4
EINECS: 231-448-7
Bereik: 01-2119489797-11
Merck Index: 12,08805

Order Now

Specification Test Results

Assay (alkalimetric) ≥99.0%
Insoluble matter ≤0.01%
pH-value (5%; water, 25 °C) 8.7 - 9.3
Chloride (Cl) ≤0.002%
Sulfate (SO₄) ≤0.005%
Heavy metals (as Pb) ≤0.001%
Cu (Copper) ≤0.0003%
Fe (Iron) ≤0.001%
K (Potassium) ≤0.01%
Pb (Lead) ≤0.001%
Loss on Drying (105 °C) ≤0.2%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR