Natriumsulfaat decahydraat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

1.06648.1000 1.06648.9025
1.06648.1000EA 98 EUR
1.06648.1000 1.06648.9025
Natriumsulfaat decahydraat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Natriumsulfaat 10 H2O
Formule: Na₂SO₄·10H₂O
MW: 322.2 g/mol
Kookpunt: 1429
Smeltpunt: 32.4
Dichtheid: 1.464 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149189
CAS nummer: 7727-73-3
EINECS: 231-820-9
Merck Index: 13,08755

Order Now

Specification Test Results

Solubility 420 g/l (20 °C)
Melting Point 32.4 °C
Molar Mass 322.19 g/mol
Bulk Density 700 kg/m3
Density 1.464 g/cm3
pH 5 - 7.5 (100 g/l, H2O, 20 °C) (anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR