Waterstofperoxide 30% Stabilised, (Perhydrol®) EMSURE® ISO analysis_for, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


MQ300

1.07210.0250 1.07210.1000 1.07210.2500 1.07210.9025
1.07210.0250EA 49.8 EUR
1.07210.0250 1.07210.1000 1.07210.2500 1.07210.9025
Waterstofperoxide 30% Stabilised, (Perhydrol®) EMSURE® ISO analysis_for, Supelco®
Waterstofperoxide
Formule: H₂O₂
MW: 34.01 g/mol
Kookpunt: ∼107 °C (1013 hPa)
Smeltpunt: –25.7 °C
Dichtheid: 1.11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011333
CAS nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8) II
Merck Index: 14,04798

Order Now

Specification Test Results

Assay (Manganometric) 30% min.
Color 10 Hazen max.
Other specifications on request 107 °C (1013 hPa)
Vapor Pressure 18 hPa (20 °C)
Density 1.11 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR