Fenolftaleïne 1% in ethanol, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

M-Clarity™ quality level = MQ200

1.07227.0250 1.07227.1000
1.07227.0250EA 65.6 EUR
1.07227.0250 1.07227.1000
Fenolftaleïne 1% in ethanol, Millipore®
Fenolftaleïne
Formule: C₂₀H₁₄O₄
MW: 318.33 g/mol
Kookpunt: ∼78 °C (1013 hPa)
Dichtheid: ∼0.85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005913
CAS nummer: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 III
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

indicator pH 8.2 - 9.8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR