Lead(II) oxide analytical reagent, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Lead monoxide

M-Clarity™ quality level = MQ300

1.07401.0250 1.07401.1000
1.07401.0250EA 211 EUR
1.07401.0250 1.07401.1000
Lead(II) oxide analytical reagent, Supelco®
Lead(II) oxide
Formule: PbO
MW: 223.2 g/mol
Kookpunt: 1470
Smeltpunt: 888
Dichtheid: 9.5 g/cm³ (20 °C)
MDL Nummer: MFCD00011164
CAS nummer: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9 III
Merck Index: 13,05431

Order Now

Specification Test Results

Solubility 0.017 g/l (20 °C)
Melting Point 888 °C
Molar Mass 223.19 g/mol
Bulk Density 3500 - 3700 kg/m3
Boiling Point 1470 °C
Density 9.53 g/cm3 (20 °C)
pH 8 - 9 (100 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR