Sand (sea sand) analysis_for, purified_acid, calcinated, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


MQ200

1.07712.1000 1.07712.5000 1.07712.9025 1.07712.9010
1.07712.1000EA 143 EUR
1.07712.1000 1.07712.5000 1.07712.9025 1.07712.9010
Sand (sea sand) analysis_for, purified_acid, calcinated, Supelco®
Sand (sea sand)
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: >2200 °C (1013 hPa)
Smeltpunt: 1713 °C
Dichtheid: 2.65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 14808-60-7
EINECS: 231-545-4

Order Now

Specification Test Results

Substances soluble in hydrochloric acid ≤ 0.2 %
Chloride (Cl) ≤ 0.005 %
Acid-soluble Iron (Fe) ≤ 0.01 %
Loss on ignition (900 °C) ≤ 0.05 %
Particle size (0.1 - 0.315 mm) ≥ 90 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR