Selenium, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ300

1.07714.0050 1.07714.0250 1.07714.1000
1.07714.0050EA 164 EUR
1.07714.0050 1.07714.0250 1.07714.1000
Selenium, Supelco®
Selenium
Formule: Se
Kookpunt: ∼685 °C (1013 hPa)
Smeltpunt: 217 °C
Dichtheid: 4,28 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134090
CAS nummer: 7782-49-2
EINECS: 231-957-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,08572

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 217 °C
Molar Mass 78.96 g/mol
Bulk Density 1270 kg/m3
Boiling Point 685 °C (1013 hPa)
Vapor Pressure 0.001 hPa (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR