Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.08045.0001
1.08045.0001EA 77.5 EUR
1.08045.0001
Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck
Milieutestkits
This refill pack contains all the reagents for the MQuant® test kit 1.18765 for the colorimetric evaluation of silicate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical system.
  • Colorimetric
  • Microbialanalysis
The test can be used for the determination of silicate e.g. in groundwater and surface water, drinking water and industrial water.