L(+)-Kaliumnatriumtartraat tetrahydraat, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

M-Clarity™ quality level = MQ300

1.08087.0500 1.08087.1000 1.08087.5000 1.08087.9050 1.08087.9012
1.08087.0500EA 90.6 EUR
1.08087.0500 1.08087.1000 1.08087.5000 1.08087.9050 1.08087.9012
L(+)-Kaliumnatriumtartraat tetrahydraat, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®
L(+)-Kaliumnatriumtartraat tetrahydraat
Formule: KOOC(CHOH)₂COONa·4H₂O
Smeltpunt: 70…80 °C
Dichtheid: ∼1,767 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150989
CAS nummer: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 630 g/l (20 °C)
Melting Point 70 - 80 °C
Molar Mass 282.23 g/mol
Bulk Density 1000 kg/m3
Density 7.0 - 8.5 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR