Tris-(hydroxymethyl)-aminomethaan (TRIS), EMPROVE® EXPERT BP, Ph. Eur., ACS, JP, USP, high purity for biopharmaceutical production

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

1.08307.9025 1.08307.1000
1.08307.9025EA 0 EUR
1.08307.9025 1.08307.1000
Tris-(hydroxymethyl)-aminomethaan (TRIS), EMPROVE® EXPERT BP, Ph. Eur., ACS, JP, USP, high purity for biopharmaceutical production
Tris-(hydroxymethyl)-aminomethaan (TRIS)
Formule: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Kookpunt: 357
Smeltpunt: 169…172 °C
Dichtheid: 1.328 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR