Tolueen, LiChrosolv® HPLC, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Gevaar
Synoniemen: Methylbenzene, Phenylmethane, FM approval
MQ100
1.08327.1000 1.08327.4000 1.08327.2500
1.08327.1000EA 97.7 EUR
1.08327.1000 1.08327.4000 1.08327.2500
Tolueen, LiChrosolv® HPLC, Supelco®
Tolueen
Formule: C₆H₅CH₃
MW: 92.14 g/mol
Kookpunt: 110.6
Smeltpunt: –95 °C
Dichtheid: 0.867 g/cm³ (20 °C)
Vlampunt: 4
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
Bereik: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Solubility 0.52 g/l (20 °C)
Melting Point -95 °C
Molar Mass 92.14 g/mol
Boiling Point 110.6 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.4968 (20 °C)
Explosion Limit 1.2 - 8 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 535 °C
Viscosity kinematic 0.7 mm2/s (20 °C)
Synonyms Toluol, Methylbenzene
Hill Formula C7H8
Chemical Formula C6H5CH3