3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
Synoniemen: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
M-Clarity™ quality level = MQ100
1.08475.1000
1.08475.1000EA 81.6 EUR
1.08475.1000
3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
3,3',5,5'-Tetramethylbenzidine (TMB)
Formule: C₁₆H₂₀N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007748
CAS nummer: 54827-17-7
UN: 3264
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Density 1.023 g/cm3
Hazard Class 8
Hazard Class Description Corrosive material
For Detection of chlorine