Jood 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

1.09099.1000 1.09099.1003
1.09099.1000EA 40.43 EUR
1.09099.1000 1.09099.1003
Jood 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Jood

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formule: I₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011355
CAS nummer: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR