Kaliumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Potash caustic, Caustic potash

M-Clarity™ quality level = MQ100

1.09112.1000
1.09112.1000EA 43.7 EUR
1.09112.1000
Kaliumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Kaliumhydroxide
Formule: KOH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR