Azure II voor microscopie (C.I.52010/52015)

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ400

1.09211.0010 1.09211.1000
1.09211.0010EA 50.3 EUR
1.09211.0010 1.09211.1000
Azure II voor microscopie (C.I.52010/52015)
Azure II

Azure II (C.I. 52010/ 52015) is a dry dye for the preparation of a color staining solution used for the staining of blood and bone marrow smears of human origin. The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.


  • Blood smears

Formule: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00080716
CAS nummer: 37247-10-2
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR