Zoutzuur concentrate for dilution to 1 mol/l in aqueous solution, Titrisol®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09970.0001 1.09970.0050
1.09970.0001EA 23.92 EUR
1.09970.0001 1.09970.0050
Zoutzuur concentrate for dilution to 1 mol/l in aqueous solution, Titrisol®, Supelco®
Zoutzuur
Formule: HCl
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.09 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR