Zwavelzuur 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

1.09984.0001 1.09984.0050
1.09984.0001EA 22.85 EUR
1.09984.0001 1.09984.0050
Zwavelzuur 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Zwavelzuur
Formule: H₂SO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.06 g/cm3 (20 °C)
pH 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR