Dithiothreitol (DTT, Cleland's reagent) voor de biochemie, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

1.11474.0001 1.11474.0005 1.11474.0025 1.11474.0500
1.11474.0001EA 48.6 EUR
1.11474.0001 1.11474.0005 1.11474.0025 1.11474.0500
Dithiothreitol (DTT, Cleland's reagent) voor de biochemie, Millipore®
Dithiothreitol (DTT, Cleland's reagent)
Formule: C₄H₁₀O₂S₂
Kookpunt: ∼125 °C (1013 hPa)
Smeltpunt: 42…44 °C
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004877
CAS nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR