Kaliumhydroxide 2 mol/l (2 N) methanol, Titripur® Volumetric, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potash caustic, Caustic potash

MQ100

1.11787.2500
1.11787.2500EA 129 EUR
1.11787.2500
Kaliumhydroxide 2 mol/l (2 N) methanol, Titripur® Volumetric, Supelco®
Kaliumhydroxide
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 1320
Smeltpunt: 360
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3 (6.1, 8) II
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR