Azomethine H sodium salt, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Synoniemen: Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt, Azomethine H, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid monosodium salt, Azomethine-H monosodium salt
MQ100
1.11962.0010 1.11962.0100
1.11962.0010EA 648 EUR
1.11962.0010 1.11962.0100
Azomethine H sodium salt, Supelco®
Azomethine H sodium salt
Formule: C₁₇H₁₂NNaO₈S₂
MW: 445.41 g/mol
Smeltpunt: > 200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042050
CAS nummer: 5941-07-1
EINECS: 227-698-1
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Molar Mass 445.39 g/mol
Bulk Density 500 kg/m3