Lithium metaborate, Spectromelt® A20 XRF, fused, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


MQ100

1.12996.1000
1.12996.1000EA 910 EUR
1.12996.1000
Lithium metaborate, Spectromelt® A20 XRF, fused, Supelco®
Lithium metaborate
Formule: BLiO₂
MW: 49.75 g/mol
Kookpunt: 1719
Smeltpunt: 845
Dichtheid: 1.4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011089
CAS nummer: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR