Natriumdodecylsulfaat, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

M-Clarity™ quality level = MQ100

1.13760.0100 1.13760.1000 1.13760.2500 1.13760.9025
1.13760.0100EA 85.8 EUR
1.13760.0100 1.13760.1000 1.13760.2500 1.13760.9025
Natriumdodecylsulfaat, Millipore®
Natriumdodecylsulfaat
Formule: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Smeltpunt: 204…207 °C
Dichtheid: 1.03 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036175
CAS nummer: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR