Preparative sorbents, LiChroprep®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
1.09303.0100EA 0 EUR
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
Preparative sorbents, LiChroprep®
Sorbenten voor chromatografie Sorbenten voor HPLC
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.