Palladium standard solution, 1000 mg/L Pd in 0.5 M nitric acid (from Pd(NO₃)₂) Certipur® standard for AAS, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.14282.0100 1.14282.0500
1.14282.0100EA 67.3 EUR
1.14282.0100 1.14282.0500
Palladium standard solution, 1000 mg/L Pd in 0.5 M nitric acid (from Pd(NO₃)₂) Certipur® standard for AAS, Supelco®
Standaarden Kationische standaarden Palladium standaarden
Certificaten: Traceable to SRM from NIST
Order Now
Specificaties
Environmentally Preferable
M-Clarity quality level MQ100